3-Bromo-4-chloro-2-nitroaniline

Leverantör: Apollo Scientific
Varning

OR55252-1G OR55252-250MG OR55252-5G
APOSOR55252-1GEA 16950 SEK
APOSOR55252-1G APOSOR55252-250MG APOSOR55252-5G
3-Bromo-4-chloro-2-nitroaniline
3-Bromo-4-chloro-2-nitroaniline
Formel: C₆H₄BrClN₂O₂
MW: 251.465 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD31543878
CAS nummer: 2091408-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR