4-Chloro-3-nitrobenzaldehyde 97%

Leverantör: Apollo Scientific
Varning

OR4977-10G OR4977-50G OR4977-25G OR4977-100G
APOSOR4977-10GEA 310 SEK
APOSOR4977-10G APOSOR4977-50G APOSOR4977-25G APOSOR4977-100G
4-Chloro-3-nitrobenzaldehyde 97%
4-Chloro-3-nitrobenzaldehyde
Formel: C₇H₄ClNO₃
MW: 185.57 g/mol
Smältpunkt: 61…65 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007078
CAS nummer: 16588-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR