5-Bromo-3-iodo-6-methylpyridin-2-amine 95%

Leverantör: Apollo Scientific

Synonyms: 2-Amino-5-bromo-3-iodo-6-methylpyridine

OR480261-10G OR480261-1G OR480261-5G
APOSOR480261-10GEA 7675 SEK
APOSOR480261-10G APOSOR480261-1G APOSOR480261-5G
5-Bromo-3-iodo-6-methylpyridin-2-amine 95%
5-Bromo-3-iodo-6-methylpyridin-2-amine
Formel: C₆H₆BrIN₂
MW: 312.936 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09880093
CAS nummer: 958357-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR