Methyl 6-(Isopropylamino)pyridazine-3-carboxylate

Leverantör: Apollo Scientific

OR470970-1G
APOSOR470970-1GEA 5875 SEK
APOSOR470970-1G
Methyl 6-(Isopropylamino)pyridazine-3-carboxylate
Methyl 6-(Isopropylamino)pyridazine-3-carboxylate
Formel: C₉H₁₃N₃O₂
MW: 195.218 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12152724
CAS nummer: 1184482-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR