cis-1,3-Cyclopentanedicarboxylic acid

Leverantör: Apollo Scientific
Varning

Synonyms: cis-Cyclopentane-1,3-dicarboxylic acid

OR46133-1G OR46133-5G
APOSOR46133-1GEA 360 SEK
APOSOR46133-1G APOSOR46133-5G
cis-1,3-Cyclopentanedicarboxylic acid
cis-1,3-Cyclopentanedicarboxylic acid
Formel: C₇H₁₀O₄
MW: 158.16 g/mol
Smältpunkt: 121 °C
Storage Temperature: Omgivande
MDL Number: MFCD01311175
CAS nummer: 876-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR