Dietylfumarat 95%

Leverantör: Apollo Scientific
Varning

Synonyms: Fumaric acid diethyl ester

OR4507-25G
APOSOR4507-25GEA 260 SEK
APOSOR4507-25G
Dietylfumarat 95%
Dietylfumarat
Formel: C₈H₁₂O₄
MW: 172.18 g/mol
Kokpunkt: 218…219 °C
Smältpunkt: 44228 °C
Densitet: 1.052 g/cm³ (25 °C)
Flampunkt: 92 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064455
CAS nummer: 623-91-6
EINECS: 210-819-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR