3-Methyl-isothiazole-5-sulphonyl chloride

Leverantör: Apollo Scientific

OR346292-500MG
APOSOR346292-500MGEA 0 SEK
APOSOR346292-500MG
3-Methyl-isothiazole-5-sulphonyl chloride
3-Methyl-isothiazole-5-sulphonyl chloride
Formel: C₄H₄ClNO₂S₂
MW: 197.663 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD21362487
CAS nummer: 1355334-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR