6-Chloro-2,5-dimethylpyrimidine-4-carboxylic acid ethyl ester

Leverantör: Apollo Scientific

OR346113-1G
APOSOR346113-1GEA 0 SEK
APOSOR346113-1G
6-Chloro-2,5-dimethylpyrimidine-4-carboxylic acid ethyl ester
6-Chloro-2,5-dimethylpyrimidine-4-carboxylic acid ethyl ester
Formel: C₉H₁₁ClN₂O₂
MW: 214.65 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02180838
CAS nummer: 76498-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR