N-(2-Hydroxypropyl)-2-methylprop-2-enamide

Leverantör: Apollo Scientific

Synonyms: N-(2-Hydroxypropyl)methacrylamide

OR311087-5G
APOSOR311087-5GEA 975 SEK
APOSOR311087-5G
N-(2-Hydroxypropyl)-2-methylprop-2-enamide
N-(2-Hydroxypropyl)-2-methylprop-2-enamide
Formel: C₇H₁₃NO₂
MW: 143.184 g/mol
Smältpunkt: 70 °C
Densitet: 1.002 g/cm³ (25 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00080531
CAS nummer: 21442-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR