1-Cyclopropylpiperazine

Leverantör: Apollo Scientific
Fara

OR15663-1G
APOSOR15663-1GEA 260 SEK
APOSOR15663-1G
1-Cyclopropylpiperazine
1-Cyclopropylpiperazine
Formel: C₇H₁₄N₂
MW: 126.1995 g/mol
Kokpunkt: 43 °C/7mmHg
Densitet: 0.94 g/cm³ (25 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD06254805
CAS nummer: 20327-23-5
UN: 3263
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR