Fmoc-L-Phe(4-Br)-OH 97%

Leverantör: Apollo Scientific
Varning

Synonyms: Fmoc-Phe(4-Br)-OH

OR14558-1G
APOSOR14558-1GEA 260 SEK
APOSOR14558-1G
Fmoc-L-Phe(4-Br)-OH 97%
Fmoc-L-Phe(4-Br)-OH
Formel: C₂₄H₂₀BrNO₄
MW: 466.30 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00273460
CAS nummer: 198561-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR