2-(5-Formyl-2-methoxyphenoxy)-2-phenylacetic acid

Leverantör: Apollo Scientific
Varning

OR14275-1G
APOSOR14275-1GEA 2200 SEK
APOSOR14275-1G
2-(5-Formyl-2-methoxyphenoxy)-2-phenylacetic acid
2-(5-Formyl-2-methoxyphenoxy)-2-phenylacetic acid
Formel: C₁₆H₁₄O₅
Smältpunkt: 159…161 °C
Storage Temperature: Omgivande
CAS nummer: 885949-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR