Ethyl-5-bromo-1H-indole-2-carboxylate

Leverantör: Apollo Scientific
Varning

Synonyms: Ethyl 5-bromoindole-2-carboxylate, 5-Bromoindole-2-carboxylic acid ethyl ester, Ethyl 5-Bromoindole-2-carboxylate, Ethyl 5-bromo-1H-indole-2-carboxylate

OR12095-1G
APOSOR12095-1GEA 260 SEK
APOSOR12095-1G
Ethyl-5-bromo-1H-indole-2-carboxylate
Ethyl-5-bromo-1H-indole-2-carboxylate
Formel: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Smältpunkt: 163…167 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00022701
CAS nummer: 16732-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR