2-Acetyl-1-naphthol

Leverantör: Apollo Scientific

Synonyms: 1'-Hydroxy-2'-acetonaphthone, 1-(1-Hydroxynaphthalen-2-yl)ethan-1-one, 2-Acetyl-1-hydroxynaphthalene, 1-Hydroxy-2-acetonaphthone

OR1123-100G OR1123-25G
APOSOR1123-100GEA 3675 SEK
APOSOR1123-100G APOSOR1123-25G
2-Acetyl-1-naphthol
2-Acetyl-1-naphthol
Formel: C₁₂H₁₀O₂
MW: 186.21 g/mol
Smältpunkt: 98…100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003963
CAS nummer: 711-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR