1-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]piperidine-4-carboxylic acid

Leverantör: Apollo Scientific

OR110879-1G
APOSOR110879-1GEA 4375 SEK
APOSOR110879-1G
1-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]piperidine-4-carboxylic acid
1-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]piperidine-4-carboxylic acid
Formel: C₁₀H₁₄N₂O₄S₂
MW: 290.359 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD26936323
CAS nummer: 1858255-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR