Polyetylenglykol 1.000

Leverantör: Apollo Scientific
Varning

Synonyms: PEG 1,000, PEG 900-1100

OR1064-50G
APOSOR1064-50GEA 855 SEK
APOSOR1064-50G
Polyetylenglykol 1.000
Polyetylenglykol 1.000
Formel: H(OCH₂CH₂)nOH
MW: 1000 g/mol
Smältpunkt: 61.7…66.6 °C
Densitet: 1.21 g/cm³ (25 °C)
Flampunkt: 260
Storage Temperature: Omgivande
MDL Number: MFCD00081839
CAS nummer: 25322-68-3
Merck Index: 13,07651

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR