1,4-Anhydro-D-mannitol 98

Leverantör: Apollo Scientific

BICL4260-100MG BICL4260-250MG
APOSBICL4260-100MGEA 5275 SEK
APOSBICL4260-100MG APOSBICL4260-250MG
1,4-Anhydro-D-mannitol 98
1,4-Anhydro-D-mannitol
Formel: C₆H₁₂O₅
MW: 164.156 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD14702340
CAS nummer: 7726-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR