2-Fluoro-(4-methoxycarbonyl)phenylboronic acid 98%

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: 2-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 2-Fluoro-4-methoxycarbonylphenylboronic acid

455870050
ACRO455870050EA 7825 SEK
ACRO455870050
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid 98%
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid
Formel: C₈H₈BFO₄
MW: 197.96 g/mol
Smältpunkt: 127…129 °C
Storage Temperature: Omgivande
MDL Number: MFCD08436034
CAS nummer: 603122-84-5

Order Now

Specifikation testresultat

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR