4-(Trifluoromethyl)styrene 98+% Stabilised

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: 1-(Trifluoromethyl)-4-vinylbenzene

Stabilization: 0.1% 4-tert-Butylbrenzcatechin

398460010
ACRO398460010EA 2400 SEK
ACRO398460010
4-(Trifluoromethyl)styrene 98+% Stabilised
4-(Trifluoromethyl)styrene

Appearance: Clear colorless to light yellow Liquid

Formel: C₉H₇F₃
MW: 172.15 g/mol
Kokpunkt: 65.8 °C (40 mmHg)
Densitet: 1.165 g/cm³
Flampunkt: 42 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00075539
CAS nummer: 402-50-6
UN: 1993
ADR: 3 III

Order Now

Specifikation testresultat

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4650 to 1.4680 (20°C, 589 nm)
GC ≥98.0 %
Stabilizer 0.1 % 4-tert.-Butylcatechol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR