Aluminiumklorid, vattenfri 99.999%, pulver for analysis

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Aluminum chloride

364810200 364810100 364811000
ACRO364810200EA 3400 SEK
ACRO364810200 ACRO364810100 ACRO364811000
Aluminiumklorid, vattenfri 99.999%, pulver for analysis
Aluminiumklorid
Formel: AlCl₃
MW: 133.34 g/mol
Kokpunkt: 180
Smältpunkt: 194 °C
Densitet: 2.44 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00003422
CAS nummer: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8 II
Merck Index: 13,00335

Order Now

Specifikation testresultat

Appearance White Powder
Total trace metal impurities ≤0.001 %
Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR