3-Methoxyphenethylamine 98+%

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: 2-(3-Methoxyphenyl)ethylamine

302610050 302610250
ACRO302610050EA 1325 SEK
ACRO302610050 ACRO302610250
3-Methoxyphenethylamine 98+%
3-Methoxyphenethylamine
Formel: C₉H₁₃NO
MW: 151.21 g/mol
Kokpunkt: 118…119 °C (6 mmHg)
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008187
CAS nummer: 2039-67-0
EINECS: 218-017-9
UN: 2735
ADR: 8 II

Order Now

Specifikation testresultat

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5375 to 1.5395 (20°C, 589 nm)
GC ≥98.0 %
Titration with HClO4 ≥98.0 %

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR