Europiumtrinitrat hexahydrat 99.9%

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Europium (III) nitrate hexahydrate

196680050
ACRO196680050EA 3050 SEK
ACRO196680050
Europiumtrinitrat hexahydrat 99.9%
Europiumtrinitrat hexahydrat
Formel: EuN₃O₉·6H₂O
MW: 446.07 g/mol
Smältpunkt: 85
MDL Number: MFCD00149700
CAS nummer: 10031-53-5
EINECS: 233-380-3
UN: 1477
ADR: 5.1 II

Order Now

Specifikation testresultat

Appearance Semitransparent Crystalline solid
Total conc. of imp.as REO ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR