Aluminiumklorid, vattenfri 99%, granulat, extra pure

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Aluminum chloride

195780010 195785000
ACRO195780010EA 895 SEK
ACRO195780010 ACRO195785000
Aluminiumklorid, vattenfri 99%, granulat, extra pure
Aluminiumklorid
Formel: AlCl₃
MW: 133.34 g/mol
Kokpunkt: 180
Smältpunkt: 194 °C
Densitet: 2.44 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00003422
CAS nummer: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8 II
Merck Index: 13,00335

Order Now

Specifikation testresultat

Appearance White to yellow to grey Granules
Titration Argentometric ≥98.5 %
Aluminium (Al) ≤150 ppm (free Al)
Chromium (Cr) ≤10 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤100 ppm
Nickel (Ni) ≤20 ppm
Zinc (Zn) ≤30 ppm
Insoluble matter ≤0.2 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR