Ammoniumacetat 98%, ren

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: Acetic acid ammonium salt

169880010 169880025 169880100
ACRO169880010EA 1275 SEK
ACRO169880010 ACRO169880025 ACRO169880100
Ammoniumacetat 98%, ren
Ammoniumacetat
Formel: H₃CCOONH₄
MW: 77.08 g/mol
Smältpunkt: 114 °C
Flampunkt: 136 °C
Storage Temperature: Omgivande
MDL Number: MFCD00013066
CAS nummer: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White Adhering crystals
Infrared spectrum Conforms
Water ≤2 %
Titration with HClO4 ≥97.5 % (On anhydrous substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR