Triphenylfosfin 99%

Leverantör: Thermo Fisher Scientific
Varning

140420010 140420250 140422500 140420050
ACRO140420010EA 3500 SEK
ACRO140420010 ACRO140420250 ACRO140422500 ACRO140420050
Triphenylfosfin 99%
Triphenylfosfin
Formel: C₁₈H₁₅P
MW: 262.29 g/mol
Kokpunkt: 377 °C (760 mmHg)
Smältpunkt: 78.5…81.5 °C
Flampunkt: 181 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003043
CAS nummer: 603-35-0
EINECS: 210-036-0
Merck Index: 12,09873

Order Now

Specifikation testresultat

Appearance White Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 78.5 to 81.5 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR