Methoxyacetonitrile 98%

Leverantör: Thermo Fisher Scientific
Varning

125660050 125660250 125661000
ACRO125660050EA 1175 SEK
ACRO125660050 ACRO125660250 ACRO125661000
Methoxyacetonitrile 98%
Methoxyacetonitrile
Formel: C₃H₅NO
MW: 71.08 g/mol
Kokpunkt: 118…119 °C (731 mmHg)
Densitet: 0.95 g/cm³
Flampunkt: 31 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001892
CAS nummer: 1738-36-9
UN: 1993
ADR: 3 III

Order Now

Specifikation testresultat

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3800 to 1.3830 (20°C, 589 nm)
GC ≥97.5 %

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR