Annexin V, FITC Labelled
Leverantör: AAT BIOQUEST
20032
AATB20032EA 4475 SEK
AATB20032
Annexin V, FITC Labelled
System för proteinlänkning och märkning
Annexins are a family of proteins that bind to phospholipid membranes in the presence of calcium.
  • Stable for six months
Quality tested in-house.
Observera: For research use only.