2,3,5-Trifenyltetrazoliumklorid

Leverantör: PanReac AppliChem
Varning

Synonyms: Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC

A6246.0025
A6246.0025EA 2550 SEK
A6246.0025
2,3,5-Trifenyltetrazoliumklorid
2,3,5-Trifenyltetrazoliumklorid
Formel: C₁₉H₁₅ClN₄
MW: 334.81 g/mol
Kokpunkt: ∼360 °C (1013 hPa)
Smältpunkt: 235
Storage Temperature: Kylskåp
MDL Number: MFCD00011963
CAS nummer: 298-96-4
EINECS: 206-071-6
Merck Index: 13,09815

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR