Mangansulfat monohydrat ≥99%

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Manganese sulphate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate

A17615.0E A17615.22 A17615.36
A17615.0EEA 6400 SEK
A17615.0E A17615.22 A17615.36
Mangansulfat monohydrat ≥99%
Mangansulfat monohydrat

Purity: 99% CAS No.: 10034-96-5 Molecular Formula: MnSO4.H2O Formula Weight: 169.02 (151.00anhy) Odour: Odourless pH: 3-3.5 Merck Reference: 14,5739 UN No.: 3077 MDL No.: MFCD00149159

Formel: MnSO₄·1H₂O
MW: 169.02 g/mol
Smältpunkt: 700
Densitet: 2.95
Storage Temperature: Omgivande
MDL Number: MFCD00149159
CAS nummer: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9 III
Merck Index: 13,05763

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR