ω,3-Dibromoacetophenone ≥97%

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: 2-Bromo-1-(3-bromophenyl)ethanone, 2-Bromo-1-(3-bromophenyl)-1-ethanone, α,3-Dibromoacetophenone, 2,3'-Dibromoacetophenone, 3-Bromophenacyl bromide

A15210.06 A15210.14
A15210.06EA 1875 SEK
A15210.06 A15210.14
ω,3-Dibromoacetophenone ≥97%
ω,3-Dibromoacetophenone

Purity: 97% CAS No.: 18523-22-3 Molecular Formula: C8H6Br2O Formula Weight: 277.95 Storage Temperature: 2°C to 8°C Odour: Pungent Beilstein Reference: 775786 UN No.: 3261 MDL No.: MFCD00082698

Formel: C₈H₆Br₂O
MW: 277.94 g/mol
Smältpunkt: 49…52 °C
MDL Number: MFCD00082698
CAS nummer: 18523-22-3
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR