Tri-n-butylamin 98%

Leverantör: Thermo Fisher Scientific
Fara

A14868.0D A14868.0F A14868.AP
A14868.0DEA 8750 SEK
A14868.0D A14868.0F A14868.AP
Tri-n-butylamin 98%
Tri-n-butylamin

Purity: 98% CAS No.: 102-82-9 Molecular Formula: C12H27N Formula Weight: 185.36 Flash Point: 71°(159°F) Refractive Index: 1.4290 Odour: Amine-like pH: 10.6 Melting Point: -70° Boiling Point: 216° Beilstein Reference: 1698872 Merck Reference: 14,9618 UN No.: 2542 MDL No.: MFCD00009431

Formel: C₁₂H₂₇N
MW: 185.35 g/mol
Kokpunkt: 216 °C
Smältpunkt: < –70 °C
Densitet: 0.778
Flampunkt: 71 °C (159 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009431
CAS nummer: 102-82-9
EINECS: 203-058-7
UN: 2542
ADR: 6.1 II
Merck Index: 13,09691

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR