2-[4 (2-Hydroxietyl)-1-piperazinyl]-ethan sulfonsyra 99

Leverantör: Thermo Fisher Scientific

Synonyms: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

A14777.30 A14777.09 A14777.18
A14777.30EA 5075 SEK
A14777.30 A14777.09 A14777.18
2-[4 (2-Hydroxietyl)-1-piperazinyl]-ethan sulfonsyra 99
HEPES

Purity: 99% CAS No.: 7365-45-9 Molecular Formula: C8H18N2O4S Formula Weight: 238.31 Physical Form: Powder Appearance: White Melting Point: ca 236° dec. Beilstein Reference: 883043 Merck Reference: 14,4654 MDL No.: MFCD00006158


A zwitterionic buffer

Formel: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
Smältpunkt: ∼ 236 °C (dec.)
Densitet: 1.448 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006158
CAS nummer: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR