1,2,4-Trihydroxibenzen ≥97%

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: Benzene-1,2,4-triol, 1,2,4-Benzenetriol

A13069.03 A13069.06 A13069.14
A13069.03EA 920 SEK
A13069.03 A13069.06 A13069.14
1,2,4-Trihydroxibenzen ≥97%
1,2,4-Trihydroxibenzen

Purity: 97% CAS No.: 533-73-3 Molecular Formula: C6H6O3 Formula Weight: 126.11 Melting Point: ca 140° subl. Beilstein Reference: 2042863 Merck Reference: 14,1073 MDL No.: MFCD00002198

Formel: C₆H₆O₃
MW: 126.11 g/mol
Kokpunkt: 362
Smältpunkt: ∼ 140 °C (subl.)
Densitet: 1.45 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002198
CAS nummer: 533-73-3
EINECS: 208-575-1
Merck Index: 13,01073

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR