Bariumnitrat ≥99%

Leverantör: Thermo Fisher Scientific
Varning

A11305.36 A11305.0E
A11305.36EA 755 SEK
A11305.36 A11305.0E
Bariumnitrat ≥99%
Barium nitrate

Purity: 99% CAS No.: 10022-31-8 Molecular Formula: BaN2O6 Formula Weight: 261.35 Melting Point: 592° dec. Merck Reference: 14,983 MDL No.: MFCD00003442

Formel: Ba(NO₃)₂
MW: 261.34 g/mol
Smältpunkt: 592 °C (dec.)
Densitet: 3.24
Storage Temperature: Omgivande
MDL Number: MFCD00003442
CAS nummer: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1 II
Merck Index: 13,00986

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR