ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])

Leverantör: PanReac AppliChem
Varning

Synonyms: ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt

A1088.0005 A1088.0010
A1088.0005EA 4425 SEK
A1088.0005 A1088.0010
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Formel: C₁₈H₂₄N₆O₆S₄
MW: 548.68 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00010404
CAS nummer: 30931-67-0
EINECS: 250-396-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR