1,1,1-Triklor-2-metyl-2-propanol hemihydrat ≥98%

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: 1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate, Chlorbutanol hemihydrate, Chlorobutanol hemihydrate, Chlorobutanol hydrous

A10181.14 A10181.22
A10181.14EA 800 SEK
A10181.14 A10181.22
1,1,1-Triklor-2-metyl-2-propanol hemihydrat ≥98%
1,1,1-Triklor-2-metyl-2-propanol hemihydrat

Purity: 98% CAS No.: 6001-64-5 Molecular Formula: C4H7Cl3O.0.5H2O Formula Weight: 186.47 (177.46anhy) Flash Point: 100°(212°F) Melting Point: 98-100°(anhy) Boiling Point: 167° Beilstein Reference: 878167 Merck Reference: 14,2129 MDL No.: MFCD00004461

Formel: C₄H₇Cl₃O·0.5H₂O
MW: 372.93 g/mol
Kokpunkt: 167 °C
Smältpunkt: 98…100 °C (anhydr.)
Flampunkt: >100 °C (212 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004461
CAS nummer: 6001-64-5
EINECS: 200-317-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR