Kolesterol

Leverantör: PanReac AppliChem

Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol

A0807.0100
A0807.0100EA 1675 SEK
A0807.0100
Kolesterol
Kolesterol
Formel: C₂₇H₄₆O
MW: 386.66 g/mol
Kokpunkt: 360
Smältpunkt: 147…150 °C
Densitet: 1.07 g/cm³ (20 °C)
Storage Temperature: Frys
MDL Number: MFCD00003646
CAS nummer: 57-88-5
EINECS: 200-353-2
Merck Index: 13,02221

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR