VWR®, Cold Cranking Simulator (CCS) viscosity standards
Leverantör: VWR Chemicals
High quality CCS viscosity standards are ideal for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing.
- Full traceability to national standards
- Ensures full compliance to ASTM and IP test method protocols
- Tamper-evident security packaging
- CCS kits available (set of 8, 14 or 18)
These Cold Cranking Simulator (CCS) viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
Certifikat: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.
Observera: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.
SPECIFIKATIONER
Environmentally Preferable |