VWR®, Cold Cranking Simulator (CCS) viscosity standards
Leverantör: VWR Chemicals
99401.260EA 1225 SEK
99401.260 99402.260 99403.001 99404.001 99405.001 99376.260 99377.260 99378.260 99379.260 99380.260 99381.260 99382.260 99383.260 99384.260 99385.260 99386.260 99387.260 99388.260 99389.260 99390.260 99391.260 99392.260 99393.260 99394.260 99395.260 99396.260 99397.260 99398.260 99399.260 99400.260
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Standarder Standarder för Viskosimetri
High quality CCS viscosity standards are ideal for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing.
  • Full traceability to national standards
  • Ensures full compliance to ASTM and IP test method protocols
  • Tamper-evident security packaging
  • CCS kits available (set of 8, 14 or 18)
These Cold Cranking Simulator (CCS) viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
Certifikat: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.
Observera: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.