VWR®, Multi-Parameter Refractive Index and Density Certified Reference Material At 15, 20 and 25 °C
Leverantör: VWR Chemicals
99173.030EA 3400 SEK
99173.030 99174.030 99175.030 99176.030
VWR®, Multi-Parameter Refractive Index and Density Certified Reference Material At 15, 20 and 25 °C
Standarder Standarder för brytningsindex
The Refractive Index and Density Certified Reference Materials provide a flexible, cost-effective and efficient solution to any laboratory calibrating and/or verifying refractive index and density measurements – all from a single material.
  • Density certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols
  • Low levels of uncertainty, ensuring maximum accuracy of data and dependable results
  • Supplied in 30 ml sealed glass vials for ample measurement readings
  • 12 month shelf life
In particular, Refractive Index and Density Certified Reference Materials are of significance to technicians with combined density and refractive index instruments to calibrate/verify their equipment’s performance and help laboratory managers meet their quality assurance requirements.
Certifikat: UKAS ISO 17025 & ISO 17034.
Order Now
SPECIFIKATIONER
Environmentally Preferable