Ättiksyra (isättika) ≥99.9%, BAKER INSTRA-ANALYZED® för spårmetallanalys, J.T.Baker®

Leverantör: Avantor
Fara

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

9524-33EA 2050 SEK
9524-33 9524-00
Ättiksyra (isättika) ≥99.9%, BAKER INSTRA-ANALYZED® för spårmetallanalys, J.T.Baker®
Ättiksyra

Certificate of lot analysis provided.

Formel: H₃CCOOH
MW: 60.05 g/mol
Kokpunkt: 118
Smältpunkt: 17
Densitet: 1.05 g/cm³ (20 °C)
Flampunkt: 38,5
Storage Temperature: Omgivande
MDL Number: MFCD00036152
CAS nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

For Laboratory,Research,or Manufacturing Use = 99.9 %
IMPORTANT: Material will freeze if stored below 17 °C (63°F). = 0.005 %
ACS - Assay (by GC, corrected for water) ≥ 99.9 %
ACS - Acetaldehyde ≤ 0.005 %
ACS - Acetic Anhydride ((CH₃CO)₂O) ≤ 0.01 %
ACS - Color (APHA) ≤ 10
Dilution Test Passes Test
ACS - Residue after Evaporation ≤ 5 ppm
ACS - Solubility in H₂O Passes Test
ACS - Specific Gravity at 20°/20°C ≥ 1.048
ACS - Substances Reducing Dichromate Passes Test
ACS - Substances Reducing Permanganate Passes Test
ACS - Titrable Base (meq/g) ≤ 0.0004
Chloride (Cl) ≤ 0.5 ppm
Phosphate (PO₄) ≤ 1 ppm
Sulfate (SO₄) ≤ 0.5 ppm
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Arsenic and Antimony (as As) ≤ 5.0 ppb
Trace Impurities - Barium (Ba) ≤ 20.0 ppb
Trace Impurities - Beryllium (Be) ≤ 10.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 50 ppb
Trace Impurities - Boron (B) ≤ 10.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 5.0 ppb
Trace Impurities - Calcium (Ca) ≤ 100.0 ppb
Trace Impurities - Chromium (Cr) ≤ 50 ppb
Trace Impurities - Cobalt (Co) ≤ 10.0 ppb
Trace Impurities - Copper (Cu) ≤ 10.0 ppb
Trace Impurities - Germanium (Ge) ≤ 50.0 ppb
Heavy Metals (as Pb) ≤ 300.0 ppb
Trace Impurities - Iron (Fe) ≤ 50.0 ppb
Trace Impurities - Lead (Pb) ≤ 10.0 ppb
Trace Impurities - Lithium (Li) ≤ 50.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 50.0 ppb
Trace Impurities - Manganese (Mn) ≤ 5.0 ppb
Trace Impurities - Mercury (Hg) ≤ 5.0 ppb
Trace Impurities - Nickel (Ni) ≤ 10.0 ppb
Trace Impurities - Potassium (K) ≤ 200.0 ppb
Trace Impurities - Silicon (Si) ≤ 50 ppb
Trace Impurities - Silver (Ag) ≤ 5.0 ppb
Trace Impurities - Sodium (Na) ≤ 500.0 ppb
Trace Impurities - Strontium (Sr) ≤ 5.0 ppb
Trace Impurities - Tin (Sn) ≤ 10.0 ppb
Trace Impurities - Titanium (Ti) ≤ 300.0 ppb
Trace Impurities - Zinc (Zn) ≤ 10.0 ppb

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR