Toluen ≥99.7%, BAKER ANALYZED® HPLC för HPLC/UHPLC, för spektrofotometri, J.T.Baker®

Leverantör: Avantor
Fara

Synonyms: Methylbenzene, Phenylmethane, FM approval

Packaged under nitrogen.

9351.1000EA 1350 SEK
9351.1000 9351.5000
Toluen ≥99.7%, BAKER ANALYZED® HPLC för HPLC/UHPLC, för spektrofotometri, J.T.Baker®
Toluen
Formel: C₆H₅CH₃
MW: 92.14 g/mol
Kokpunkt: 110,6
Smältpunkt: –95 °C
Densitet: 0.867 g/cm³ (20 °C)
Flampunkt: 4
Storage Temperature: Omgivande
MDL Number: MFCD00008512
CAS nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

For Laboratory,Research,or Manufacturing Use ≥ 99.7 %
Assay (by GC, corrected for water) ≥ 99.7 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤ 0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm ≤ 0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 288 nm ≤ 0.40
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤ 285
Residue after Evaporation ≤ 3.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤ 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR