Toluen ≥99.7%, ULTRA RESI-ANALYZED® för analys av organiskrester, J.T.Baker®

Leverantör: Avantor
Fara

Synonyms: Methylbenzene, Phenylmethane, FM approval

Packaged under inert gas.

9336.1000EA 1450 SEK
9336.1000 9336.2500 9336-03
Toluen ≥99.7%, ULTRA RESI-ANALYZED® för analys av organiskrester, J.T.Baker®
Toluen
Formel: C₆H₅CH₃
MW: 92.14 g/mol
Kokpunkt: 110,6
Smältpunkt: –95 °C
Densitet: 0.867 g/cm³ (20 °C)
Flampunkt: 4
Storage Temperature: Omgivande
MDL Number: MFCD00008512
CAS nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

For Laboratory,Research,or Manufacturing Use = 10
Trace Organic Residues - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤10
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
Assay (C₆H₅CH₃) (by GC, corrected for water) ≥99.7 %
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR