VWR®, Nominal flash point standard (65 °C)
Leverantör: VWR Chemicals
91688.230EA 4000 SEK
91688.230
VWR®, Nominal flash point standard (65 °C)
Standarder Petroleum Industry Standards
Observera: Carcinogen.
Order Now
SPECIFIKATIONER
Benämning Nominal Flash Point Standard (65°C)
Environmentally Preferable
CAS-nr 1120-21-4
Formel C₁₁H₂₄
Förvaringstemperatur Ambient (>5 °C)