Molybdensyra 85 zS

Leverantör: VWR Chemicals

91302.180EA 1100 SEK
91302.180
Molybdensyra 85 zS
Molybdensyra
Formel: H₂MoO₄
MW: 161.95 g/mol
Kokpunkt: ∼300 °C (1013 hPa)
Smältpunkt: 300 °C
Densitet: 3.1 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00036287
CAS nummer: 7782-91-4
EINECS: 231-970-5
Merck Index: 13,06262

Order Now

Specifikation testresultat

Assay Min 85%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR