VWR®, Mixed Anions Standard, 7 components in H₂O
Leverantör: VWR Chemicals
91295.180EA 2500 SEK
91295.180
VWR®, Mixed Anions Standard, 7 components in H₂O
Standarder Anionic Standards Multi-Element Anioinic Standards
7 components: 100 mg/l each of Chlorides (Cl-), Fluorides (F-), Sulphates (SO42-), Nitrates (NO3-), Nitrites (NO2-), Bromides (Br-), Phosphates (PO43-) in H₂O
Certifikat: ISO 17025/17034
Order Now
SPECIFIKATIONER
Environmentally Preferable
Förvaringstemperatur Ambient