Preparative HPLC system, Prep LC Premier
Leverantör: KNAUER
PREPLCSET2 PREPLCSET3 PREPLCSET4 PREPLCSET5
905-0698EA 0 SEK
905-0698 905-0699 905-0700 906-0701
Preparative HPLC system, Prep LC Premier
Kromatografisystem HPLC system, preparativt
The Knauer Prep LC Premier LC system is a very flexible preparative chromatography system to optimise your purification and synthesis work. The modular system can be perfectly adapted to your need, whatever kind of application you want to use. Usually it's been used with special edition of Dataapex Clarity (Clarity Prep), perfect for preparative work in a modern lab. To give you a small overview of the potential of the Prep LC Premier, we created 5 Prep bundles. These packages are the first step to build your individualised Prep LC Premier system.
  • Modular system
  • Low and High pressure
More Product Information