Cyanine 488NS succinimidyl ester dye_cyanine, sulphonated_non
Leverantör: Biotium
90117-1MG
90117-1MGEA 2375 SEK
90117-1MG
Cyanine 488NS succinimidyl ester dye_cyanine, sulphonated_non
Cyanine succinimidyl ester
Cyanine NS succinimidyl esters are non-sulfonated cyanine dyes that can be used to label proteins, oligonucleotides and nucleic acids. Available in 3 colors, Cyanine 488NS, Cyanine 555NS, and Cyanine 647NS are designed to match by charge and molecular weight for 2D DIGE platform.
  • Ex/Em 483/508 nm
  • MW added to protein: 437
  • Orange solid soluble in DMF or DMSO
  • Store desiccated at -20°C; protect from light
Cyanine NS succinimidyl esters are non-sulfonated cyanine dyes that can be used to label proteins, oligonucleotides and nucleic acids. Available in 3 colors, Cyanine 488NS, Cyanine 555NS and Cyanine 647NS are non-sulfonated versions of Cy®2, Cy®3, and Cy®5, respectively. They are designed to match by charge and molecular weight for 2D DIGE platform. Biotium offers these dyes as high quality amine-reactive succinimidyl esters.
Also see sulfonated Cyanine Dye Succinimidyl Esters and our CF® Dye Succinimidyl Esters, available with more than 20 bright and photostable CF® Dyes.
Cy2, Cy3, Cy5 are registered trademarks of GE Healthcare.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b35e47b5f1804bbf42d7280894f96d2889e369c&vid=dab94bcdf91ee56f3d0bfadda9ecb856276ca0b5&prid=web&clid=SAPDAM