di-natriumoxalat concentrate for dilution to 0.1 N i vattenlösning, Acculute™ standard volumetric solution

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances

88626.LQ 88626.LS
88626.LQEA 1900 SEK
88626.LQ 88626.LS
di-natriumoxalat concentrate for dilution to 0.1 N i vattenlösning, Acculute™ standard volumetric solution
di-natriumoxalat

CAS No.: 62-76-0 Molecular Formula: Na2C2O4 Formula Weight: 134.00 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00012465

Formel: Na₂C₂O₄
MW: 134 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00012465
CAS nummer: 62-76-0
EINECS: 200-550-3
Merck Index: 13,08723

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR