Aluminiumklorid, vattenfri ≥99.985%

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Aluminum chloride

88488.06 88488.18 88488.30
88488.06EA 2075 SEK
88488.06 88488.18 88488.30
Aluminiumklorid, vattenfri ≥99.985%
Aluminiumklorid

Purity: 99.985% CAS No.: 7446-70-0 Molecular Formula: AlCl3 Formula Weight: 133.34 Physical Form: Powder Density: 2.44 Appearance: White to yellow Odour: Pungent pH: 2,4 Melting Point: 190° subl. Merck Reference: 14,337 UN No.: 1726 MDL No.: MFCD00003422

Formel: AlCl₃
MW: 133.34 g/mol
Kokpunkt: 180
Smältpunkt: 190 °C (subl.)
Densitet: 2.44
Storage Temperature: Omgivande
MDL Number: MFCD00003422
CAS nummer: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8 II
Merck Index: 14,337

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR