Barium titanate 99,7 Metaller basis

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

88267.22 88267.36
88267.22EA 2225 SEK
88267.22 88267.36
Barium titanate 99,7 Metaller basis
Barium titanate

Purity: 99.7% CAS No.: 12047-27-7 Molecular Formula: BaTiO3 Formula Weight: 233.19 Physical Form: 2 Micron Powder Density: 5.85 Appearance: White Odour: Odourless Melting Point: 1620° Merck Reference: 14,1000 MDL No.: MFCD00003447

Formel: BaO₃Ti
MW: 233.21 g/mol
Smältpunkt: 1620 °C
Densitet: 5.85
Storage Temperature: Omgivande
MDL Number: MFCD00003447
CAS nummer: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1 III
Merck Index: 14,1000

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR